Al-23 Tube (one end closed);OD (mm), 4;ID (mm), 2
Thermo Scientific Chemicals
Al-23 Tube (one end closed);OD (mm), 4;ID (mm), 2
数量:
200 mm
300 mm
600 mm
800 mm
1000 mm
製品番号(カタログ番号) 030062.DR
または、製品番号030062-DR
価格(JPY)
-
数量:
200 mm
一括またはカスタム形式をリクエストする
仕様
化学物質名または材質Al-23 Tube
名称ノートOne end closed
推奨保存方法Ambient temperatures
TSCAU
数量200 mm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
よくあるご質問(FAQ)
What is the maximum working temperature of Al-23 Tubes?
The maximum working temperature for the Al-23 Tube is 1950 degrees C.