Carnauba wax
Thermo Scientific Chemicals

Carnauba wax

CAS: 8015-86-9
数量:
250 g
1 kg
5 kg
製品番号(カタログ番号) 031117.A7
または、製品番号031117-A7
価格(JPY)
-
数量:
5 kg
一括またはカスタム形式をリクエストする
仕様
CAS8015-86-9
化学物質名または材質Carnauba wax
密度0.990g/mL to 0.999g/mL
EINECS番号232-399-4
MDL番号MFCD00130724
さらに表示
Carnauba wax is used as a glazing agent, bulking agent, acidity regulator and tablet-coating agent. It is also used in automobile waxes, shoe polishes, dental floss and food products such as sweets, instrument polishes, floor, furniture waxes, polishes and paper coatings. It serves as the main ingredient in surfboard wax. Further, it is also used as a mold release agent to produce fiber-reinforced plastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only