Iron(II) bromide hydrate
Iron(II) bromide hydrate
Iron(II) bromide hydrate
Thermo Scientific Chemicals

Iron(II) bromide hydrate

CAS: 13463-12-2 | Br2Fe | 215.65 g/mol
製品番号(カタログ番号) 031133.14
または、製品番号031133-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13463-12-2
Assay (unspecified)Br 52% min
It is widely as a pharmaceutical intermediate.It is a useful synthetic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only