Sodium bis(trimethylsilyl)amide, 98%
Sodium bis(trimethylsilyl)amide, 98%
Sodium bis(trimethylsilyl)amide, 98%
Thermo Scientific Chemicals

Sodium bis(trimethylsilyl)amide, 98%

CAS: 1070-89-9 | C6H18NNaSi2 | 183.377 g/mol
製品番号(カタログ番号) 033505.09
または、製品番号033505-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1070-89-9
IUPAC Namesodium bis(trimethylsilyl)azanide
Molecular FormulaC6H18NNaSi2
InChI KeyWRIKHQLVHPKCJU-UHFFFAOYSA-N
SMILES[Na+].C[Si](C)(C)[N-][Si](C)(C)C
さらに表示
Appearance (Color)White to pale cream or pale yellow
FormPowder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting point171-175°C
Sodium bis(trimethylsilyl)amide is used as a base in organic synthesis as well as to deprotonate other compounds containing weakly acidic O-H, S-H, and N-H bonds. It is used to prepare amines by reacting with alkyl halides. It finds application in certain organic reactions viz. generation of carbenes by dehydrohalogenation of halocarbons and Wittig reagents by deprotonation of phosphonium salts. It accelerates the polymerization of pheneylacetylene in conjunction with rhodium(I) catalysis. Furthermore, it is used for the preparation of lanthanide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only