Strontium bromide, ultra dry, 99.995% (metals basis)
Strontium bromide, ultra dry, 99.995% (metals basis)
Strontium bromide, ultra dry, 99.995% (metals basis)
Thermo Scientific Chemicals

Strontium bromide, ultra dry, 99.995% (metals basis)

CAS: 10476-81-0 | Br2Sr | 247.428 g/mol
製品番号(カタログ番号) 035717.03
または、製品番号035717-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10476-81-0
FormPowder
Assay (unspecified)Br 63.5 - 65.4%
Total Metal Impurities0.005% max.
Appearance (Color)White
It is used as sedatives in medicine and analytical reagents, for facile synthesis of poly(phenylcarbyne), which is a precursor to diamond-like carbon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only