Zinc iodide, ultra dry, 99.995% (metals basis)
Zinc iodide, ultra dry, 99.995% (metals basis)
Zinc iodide, ultra dry, 99.995% (metals basis)
Thermo Scientific Chemicals

Zinc iodide, ultra dry, 99.995% (metals basis)

CAS: 10139-47-6 | ZnI2
数量:
5 g
25 g
100 g
製品番号(カタログ番号) 035727.22
または、製品番号035727-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10139-47-6
IUPAC Namezinc(2+) diiodide
Molecular FormulaI2Zn
InChI KeyUAYWVJHJZHQCIE-UHFFFAOYSA-L
SMILES[Zn++].[I-].[I-]
さらに表示
Appearance (Color)White
Total Metal Impurities0.005% max.
Residual water< 100 ppm
FormGranular
Zinc iodide is used as an x-ray opaque penetrant in industrial radiography to enhance the contrast. It is used as electrode compartments and as a stain in electron microscopy in association with osmium tetroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only