Methyl Red hydrochloride, ACS
Methyl Red hydrochloride, ACS
Methyl Red hydrochloride, ACS
Thermo Scientific Chemicals

Methyl Red hydrochloride, ACS

CAS: 63451-28-5 | C15H16ClN3O2 | 305.762 g/mol
製品番号(カタログ番号) 036668.14
または、製品番号036668-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS63451-28-5
IUPAC Namehydrogen 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid chloride
Molecular FormulaC15H16ClN3O2
InChI KeyICTWKXAVJAHRMK-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O
さらに表示
CommentClarity of alcohol solution P.T.
CommentVisual transition interval from pH 4.2 (pink) to 6.2 (yellow)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only