Iodine monobromide, 98% min
Iodine monobromide, 98% min
Iodine monobromide, 98% min
Thermo Scientific Chemicals

Iodine monobromide, 98% min

CAS: 7789-33-5 | BrI | 206.81 g/mol
製品番号(カタログ番号) 039106.22
または、製品番号039106-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7789-33-5
IUPAC Nameiodobromane
Molecular FormulaBrI
InChI KeyCBEQRNSPHCCXSH-UHFFFAOYSA-N
SMILESBrI
さらに表示
Appearance (Color)Black, grey, or blue-purple
FormCrystals or crystalline powder
Assay (unspecified)≥98.0%
Iodine bromide is used for bromination of and for determining iodine used. IBr is used in some types of iodometry. It is also used as a organic synthesis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only