Potassium 2-ethylhexanoate, 99.9% (metals basis), 75% w/w soln.
Potassium 2-ethylhexanoate, 99.9% (metals basis), 75% w/w soln.
Potassium 2-ethylhexanoate, 99.9% (metals basis), 75% w/w soln.
Thermo Scientific Chemicals

Potassium 2-ethylhexanoate, 99.9% (metals basis), 75% w/w soln.

CAS: 3164-85-0 | C8H15KO2 | 182.30 g/mol
製品番号(カタログ番号) 039163.04
または、製品番号039163-04
価格(JPY)
-
数量:
2 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3164-85-0
IUPAC Namepotassium 2-ethylhexanoate
Molecular FormulaC8H15KO2
InChI KeyZUFQCVZBBNZMKD-UHFFFAOYNA-M
SMILES[K+].CCCCC(CC)C([O-])=O
さらに表示
FormViscous liquid
Total Metal Impurities≤0.1%
Assay (unspecified)≥74.0 to ≤76.0%
Used to convert the tert-butylammmonium salt of clavulanic acid into potassium clavulanate or clavulanate potassium. Also used as a catalyst for polyurethane systems (foams) and for unsaturated polyester resin systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For Manufacturing and Laboratory Use. Not for beverage use