Cyclopentadienylmolybdenum tricarbonyl dimer, 98%
Cyclopentadienylmolybdenum tricarbonyl dimer, 98%
Cyclopentadienylmolybdenum tricarbonyl dimer, 98%
Thermo Scientific Chemicals

Cyclopentadienylmolybdenum tricarbonyl dimer, 98%

CAS: 12091-64-4 | C16H2Mo2O6 | 482.09 g/mol
製品番号(カタログ番号) 039245.03
または、製品番号039245-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12091-64-4
Appearance (Color)Red to purple
FormCrystals or crystalline powder
Assay from Supplier's CofA≥97.5%
It is widely used as a chemical intermediate in the laboratories, manufacture purposes, chemical and pharmaceutical intemediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only