Decamethylferrocene, 99%, Thermo Scientific Chemicals
Decamethylferrocene, 99%, Thermo Scientific Chemicals
Decamethylferrocene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Decamethylferrocene, 99%, Thermo Scientific Chemicals

数量:
250 mg
1 g
5 g
製品番号(カタログ番号) 039406.03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12126-50-0
Molecular FormulaC20H30Fe
InChI KeyVKRKSGODPDKGAD-UHFFFAOYSA-N
SMILESCc1c(C)c(C)[c-](C)c1C.C[C-]12[C-]3(C)[C-]4(C)[C-]5(C)[C-]1(C)[Fe]2345
Molecular Weight (g/mol)326.31
さらに表示
Assay (unspecified)> 98.5%
Appearance (Color)Yellow - orange crystals
Decamethylferrocene is used as a superior internal reference redox standard to ferrocene in voltammetric and amperometric titrations. It is used as photocathode for the detection of barium fluoride scintillation light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only