Calcium stearate
Calcium stearate
Calcium stearate
Thermo Scientific Chemicals

Calcium stearate

Waterproofing and release agent, stabilizer for PVC, lubricant, conditioning agent | CAS: 1592-23-0 | C36H70CaO4 | 607.03 g/mol
製品番号(カタログ番号) 039423.A1
または、製品番号039423-A1
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1592-23-0
IUPAC Namecalcium dioctadecanoate
Molecular FormulaC36H70CaO4
InChI KeyCJZGTCYPCWQAJB-UHFFFAOYSA-L
SMILES[Ca++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
さらに表示
CommentFree fatty acid: 0.5 % max
CommentSoftening point: 145 to 165?C
Ash9.8 to 10.8%
Loss on Drying3.5% max
Used as a stabilizer and a lubricant in rigid PVC processing, PVC compounding and moulding, in leather cloth manufacturing. It is used as a lubricant, in iron wire drawing and used as an anti caking agent in detergent Cakes and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only