Iron powder, 98% (metals basis), Thermo Scientific Chemicals
Iron powder, 98% (metals basis), Thermo Scientific Chemicals
Iron powder, 98% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron powder, 98% (metals basis), Thermo Scientific Chemicals

数量:
250 g
500 g
1 kg
製品番号(カタログ番号) 039813.36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
さらに表示
Assay from Supplier's CofAIron (Fe), metallic: ≥98.0%
FormPowder
Particle Size-45 micron: ≥80.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only