Polyethyleneimine, branched, M.W. 600, 99%
Polyethyleneimine, branched, M.W. 600, 99%
Polyethyleneimine, branched, M.W. 600, 99%
Polyethyleneimine, branched, M.W. 600, 99%
Thermo Scientific Chemicals

Polyethyleneimine, branched, M.W. 600, 99%

CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) 040527.14
または、製品番号040527-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9002-98-6
Molecular Formula((C2H5N)zC2H4N)y(C2H5N)x
SMILES*-CCNCCN(-*)CCN-*
MDL NumberMFCD00084427
Appearance (Color)Light yellow
Formclear viscous liquid
Assay (unspecified)99.0%, min
Polyethyleneimine, branched, M.W. 600 acts as an auxiliary agent. It is used in paper-making, water treatment, detergents, adhesives and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only