Palladium Thinfoil, 0.0125mm (0.0005in) thick, 99.95% (metals basis)
Palladium Thinfoil, 0.0125mm (0.0005in) thick, 99.95% (metals basis)
Palladium Thinfoil, 0.0125mm (0.0005in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Palladium Thinfoil, 0.0125mm (0.0005in) thick, 99.95% (metals basis)

CAS: 7440-05-3 | Pd | 106.42 g/mol
数量:
25 x 25 mm
50 x 50 mm
製品番号(カタログ番号) 040744.FI
または、製品番号040744-FI
価格(JPY)
-
数量:
50 x 50 mm
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
さらに表示
Appearance (Color)Silver Gray
FormFoil, 0.0125mm (0.0005in) thick.
Assay from Supplier's CofA≥99.95% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only