Chromium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Chromium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Chromium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Chromium sputtering target, 50.8mm (2.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)

CAS: 7440-47-3 | Cr | 52.00 g/mol
製品番号(カタログ番号)数量
040842.KS1 Ea.
製品番号(カタログ番号) 040842.KS
価格(JPY)
-
数量:
1 Ea.
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-47-3
IUPAC Namechromium
Molecular FormulaCr
InChI KeyVYZAMTAEIAYCRO-UHFFFAOYSA-N
SMILES[Cr]
さらに表示
Appearance (Color)Silvery gray
FormSputtering target, 50.8mm (2.0in) diameter x 3.18mm (0.125in) thick.
Assay from Supplier's CofA≥99.95% (metals basis)
Chromium sputtering target is commonly used for thin-film deposition, etching and analytical techniques. And It is also used as Magnetron sputtering source.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only