n-Pentane, Environmental Grade, 98+%
n-Pentane, Environmental Grade, 98+%
n-Pentane, Environmental Grade, 98+%
Thermo Scientific Chemicals

n-Pentane, Environmental Grade, 98+%

Suitable For Environmental, Pesticide, and GC Analysis | CAS: 109-66-0 | C5H12 | 72.15 g/mol
数量:
1 L
4 L
4 x 4 L
製品番号(カタログ番号) 040981.M6
または、製品番号040981-M6
価格(JPY)
-
数量:
4 x 4 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS109-66-0
IUPAC Namepentane
Molecular FormulaC5H12
InChI KeyOFBQJSOFQDEBGM-UHFFFAOYSA-N
SMILESCCCCC
さらに表示
Assay (GC)GC-FID ≥99.8% (C5 Hydrocarbons)
Water Content (Karl Fischer Titration)≤0.02%
Appearance (Color)Clear colorless, ≤10 APHA
Assay from Supplier's CofAGC-ECD Responsive residue (single peak) (as Heptachlorepoxide) ≤10 ppt
Residue on Evaporation≤5ppm
さらに表示
n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only