Potassium phosphate, anhydrous, 97%
Potassium phosphate, anhydrous, 97%
Potassium phosphate, anhydrous, 97%
Thermo Scientific™

Potassium phosphate, anhydrous, 97%

CAS: 7778-53-2 | K3O4P | 212.265 g/mol
数量:
250 g
1 kg
5 kg
製品番号(カタログ番号) 041587.A1
または、製品番号041587-A1
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7778-53-2
IUPAC Nametripotassium phosphate
Molecular FormulaK3O4P
InChI KeyLWIHDJKSTIGBAC-UHFFFAOYSA-K
SMILES[K+].[K+].[K+].[O-]P([O-])([O-])=O
さらに表示
Assay (unspecified)96.1% min
Potassium phosphate is used as a fertilizer and food additive. It serves as a buffering agent to regulate pH. It is also used in optical modulators and for non-linear optics such as second-harmonic generation (SHG). It is involved in the phase-transfer catalyzed preparation of acetylenes from the corresponding vicinal dibromo compounds through double dehydrobromination. It is also useful for the oxidative coupling of thiols to disulfide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only