Ethylaluminum dichloride, 1M in hexanes
Ethylaluminum dichloride, 1M in hexanes
Ethylaluminum dichloride, 1M in hexanes
Thermo Scientific Chemicals

Ethylaluminum dichloride, 1M in hexanes

CAS: 563-43-9 | C2H5AlCl2 | 126.94 g/mol
製品番号(カタログ番号) 041712.AK
または、製品番号041712-AK
価格(JPY)
-
数量:
250 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS563-43-9
IUPAC Namedichloro(ethyl)alumane
Molecular FormulaC2H5AlCl2
InChI KeyUAIZDWNSWGTKFZ-UHFFFAOYSA-L
SMILESCC[Al](Cl)Cl
さらに表示
FormLiquid
Concentration0.95-1.10M
Appearance (Color)Clear colorless to yellow
Ethylaluminum dichloride is used as a catalyst component in the polyolefin industry, for the synthesis of synthetic rubbers and for the dimerisation of olefins. It is also employed as a catalyst for Friedel-Crafts acylations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only