Polystyrene standard, M.W. 900,000, Mw/Mn 1.10
Polystyrene standard, M.W. 900,000, Mw/Mn 1.10
Polystyrene standard, M.W. 900,000, Mw/Mn 1.10
Thermo Scientific Chemicals

Polystyrene standard, M.W. 900,000, Mw/Mn 1.10

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
製品番号(カタログ番号)数量
041943.03
または、製品番号041943-03
1 g
製品番号(カタログ番号) 041943.03
または、製品番号041943-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
仕様
CAS9003-53-6
ChEBICHEBI:27452
化学物質名または材質Polystyrene standard
IUPAC名ethenylbenzene
InChIキーPPBRXRYQALVLMV-UHFFFAOYSA-N
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only