Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%
Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%
Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%
Thermo Scientific Chemicals

Potassium hydrogen iodate, primary standard, ACS, 99.95-100.05%

Maximum level of impurities: Insoluble matter 0.01%, SO4 0.005%, Fe 0.001%, Cl 0.02%, Heavy Metals (as Pb) 0.001% | CAS: 13455-24-8 | HI2KO6 | 389.909 g/mol
製品番号(カタログ番号) 041986.22
または、製品番号041986-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13455-24-8
IUPAC Namepotassium iodic acid iodate
Molecular FormulaHI2KO6
InChI KeyACAYDTMSDROWHW-UHFFFAOYSA-M
SMILES[K+].O[I](=O)=O.[O-][I](=O)=O
さらに表示
Assay (unspecified)99.95-100.05% KH(IO₃)₂
Impurity contentCl 0.02%
Impurity contentFe 0.001%
Impurity contentSO₄ 0.005%
Insoluble material0.01%
さらに表示
Potassium hydrogen iodate acts as an oxidizing agent. Further, it serves as an intermediate for the synthesis of potassium iodate and also used as an analysis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only