Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)
Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)
Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)
Thermo Scientific Chemicals

Aluminum slug, 3.175mm (0.125in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)

CAS: 7429-90-5 | Al | 26.98 g/mol
製品番号(カタログ番号) 042258.18
または、製品番号042258-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7429-90-5
IUPAC Namealuminium
Molecular FormulaAl
InChI KeyXAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES[Al]
さらに表示
Appearance (Color)Silvery white
FormSlug, 3.175mm (0.125in) diameter.
Assay from Supplier's CofA≥99.99% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only