Quartz rod, fused, 10.0mm (0.394in) dia
Quartz rod, fused, 10.0mm (0.394in) dia
Quartz rod, fused, 10.0mm (0.394in) dia
Thermo Scientific Chemicals

Quartz rod, fused, 10.0mm (0.394in) dia

CAS: 14808-60-7 | O2Si | 60.08 g/mol
数量:
12 in.
48 in.
製品番号(カタログ番号) 042294.NT
または、製品番号042294-NT
価格(JPY)
-
数量:
48 in.
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
さらに表示
CommentNo specifications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only