Titanium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.98% (metals basis), Thermo Scientific Chemicals
Titanium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.98% (metals basis), Thermo Scientific Chemicals
Titanium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.98% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Titanium slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.98% (metals basis), Thermo Scientific Chemicals

製品番号(カタログ番号)数量
042388.QK
または、製品番号042388-QK
25 Slugs
製品番号(カタログ番号) 042388.QK
または、製品番号042388-QK
価格(JPY)
-
数量:
25 Slugs
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
さらに表示
Appearance (Color)Silvery.
FormSlug, 6.35mm (0.25in) diameter.
Assay from Supplier's CofA≥99.98% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only