Xylenol Orange tetrasodium salt, 0.1% w/v aq. soln.
Xylenol Orange tetrasodium salt, 0.1% w/v aq. soln.
Xylenol Orange tetrasodium salt, 0.1% w/v aq. soln.
Thermo Scientific Chemicals

Xylenol Orange tetrasodium salt, 0.1% w/v aq. soln.

CAS: 3618-43-7 | C31H28N2Na4O13S
製品番号(カタログ番号)数量
042773.AE
または、製品番号042773-AE
100 mL
製品番号(カタログ番号) 042773.AE
または、製品番号042773-AE
価格(JPY)
-
数量:
100 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS3618-43-7
IUPAC Nametetrasodium 2-[({5-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-2,2-dioxo-3H-1,2λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxylatomethyl)amino]acetate
Molecular FormulaC31H28N2Na4O13S
InChI KeyFXYMNKLSOQONJB-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(C2=CC=CC=C2OS1(=O)=O)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1
さらに表示
Assay from Supplier's CofA0.100% w/v ± 0.005% w/v
Appearance (Color)Red
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only