Silver slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)
Silver slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)
Silver slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)
Thermo Scientific Chemicals

Silver slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol
数量:
25 Slugs
5 Slugs
製品番号(カタログ番号) 042922.QK
または、製品番号042922-QK
価格(JPY)
-
数量:
25 Slugs
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
さらに表示
Assay from Supplier's CofA≥99.99% (metals basis)
FormSlug, 6.35mm (0.25in) diameter.
Appearance (Color)Silver
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only