Bis(cyclopentadienyl)chromium, sublimed, 97+%
Bis(cyclopentadienyl)chromium, sublimed, 97+%
Bis(cyclopentadienyl)chromium, sublimed, 97+%
Thermo Scientific Chemicals

Bis(cyclopentadienyl)chromium, sublimed, 97+%

CAS: 1271-24-5 | C10H10Cr | 182.19 g/mol
製品番号(カタログ番号) 043177.03
または、製品番号043177-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1271-24-5
IUPAC NameBis(cyclopentadienyl)chromium
Molecular FormulaC10H10Cr
InChI KeyTYYBBNOTQFVVKN-UHFFFAOYSA-N
SMILES[Cr++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
さらに表示
Appearance (Color)Red to purple to black
FormCrystals or crystalline powder
Assay from Supplier's CofA≥97.0%
Provides catalyst for ethylene polymerization. A precursor to other chromium(II) compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only