Cobalt wire, 2.0mm (0.08in) dia, 99.95% (metals basis)
Cobalt wire, 2.0mm (0.08in) dia, 99.95% (metals basis)
Cobalt wire, 2.0mm (0.08in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals

Cobalt wire, 2.0mm (0.08in) dia, 99.95% (metals basis)

CAS: 7440-48-4 | Co | 58.93 g/mol
製品番号(カタログ番号) 043290.BQ
または、製品番号043290-BQ
価格(JPY)
-
数量:
5 cm
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
さらに表示
Appearance (Color)Blue silver
Assay from Supplier's CofA≥99.95% (metals basis)
FormWire, 2.0mm (0.08in) diameter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only