Aluminum oxide, catalyst support, intermediate surface area (low SiO{2})
Aluminum oxide, catalyst support, intermediate surface area (low SiO{2})
Aluminum oxide, catalyst support, intermediate surface area (low SiO{2})
Thermo Scientific Chemicals

Aluminum oxide, catalyst support, intermediate surface area (low SiO{2})

Form: S.A. 100m2/g | CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
製品番号(カタログ番号) 043857.A1
または、製品番号043857-A1
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
さらに表示
Appearance (Color)White to pale cream
FormPellets
Surface Area80-120 m²/g
CommentTotal pore volume: 0.44-0.54 cc/g
Aluminum oxide finds applications in catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs, heat-resistant fiber. Heavy quantity of aluminum oxide is used in the production of aluminum and a component of clay.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only