Silicon slugs, 9.5mm (0.375 in.) dia. x 9.5mm (0.375 in.) length, 99.9999% (metals basis), Thermo Scientific Chemicals
Silicon slugs, 9.5mm (0.375 in.) dia. x 9.5mm (0.375 in.) length, 99.9999% (metals basis), Thermo Scientific Chemicals
Silicon slugs, 9.5mm (0.375 in.) dia. x 9.5mm (0.375 in.) length, 99.9999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicon slugs, 9.5mm (0.375 in.) dia. x 9.5mm (0.375 in.) length, 99.9999% (metals basis), Thermo Scientific Chemicals

製品番号(カタログ番号)数量
043926.14
または、製品番号043926-14
25 g
製品番号(カタログ番号) 043926.14
または、製品番号043926-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-21-3
IUPAC Namesilicon
Molecular FormulaSi
InChI KeyXUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES[Si]
さらに表示
Certificate of ConformanceCertificate of Conformance Provided
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only