Dimethyl sulfoxide, anhydrous, 99.8+%, packaged under Argon in resealable ChemSeal™ bottles
Dimethyl sulfoxide, anhydrous, 99.8+%, packaged under Argon in resealable ChemSeal™ bottles
Dimethyl sulfoxide, anhydrous, 99.8+%, packaged under Argon in resealable ChemSeal™ bottles
Thermo Scientific Chemicals

Dimethyl sulfoxide, anhydrous, 99.8+%, packaged under Argon in resealable ChemSeal™ bottles

CAS: 67-68-5 | C2H6OS | 78.13 g/mol
数量:
1 L
4 x 100 mL
4 x 1 L
製品番号(カタログ番号) 043998.M1
または、製品番号043998-M1
価格(JPY)
-
数量:
4 x 1 L
一括またはカスタム形式をリクエストする
化学物質識別子
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
さらに表示
FormLiquid
Appearance (Color)Colorless
Assay (GC)≥99.8% (Corrected for water)
Residue on Evaporation≤5 ppm
CommentInert gas used can be either Argon or Nitrogen
さらに表示
Dimethyl sulfoxide is used as a solvent for chemical reactions involving salts, most notably in Finkelstein reactions and in other nucleophilic substitutions. DMSO is an effective pain relieving agent. Used in antifreeze or hydraulic fluids. Utilized in the oxidation of thiols and disulfides to sulfonic acids. Used to enhance dermal absorption of many chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only