Tricyclohexylphosphine, technical grade
Tricyclohexylphosphine, technical grade
Tricyclohexylphosphine, technical grade
Thermo Scientific Chemicals

Tricyclohexylphosphine, technical grade

CAS: 2622-14-2 | C18H33P | 280.436 g/mol
製品番号(カタログ番号) 044854.03
または、製品番号044854-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2622-14-2
IUPAC Nametricyclohexylphosphane
Molecular FormulaC18H33P
InChI KeyWLPUWLXVBWGYMZ-UHFFFAOYSA-N
SMILESC1CCC(CC1)P(C1CCCCC1)C1CCCCC1
さらに表示
Appearance (Color)White to off-white
Formsolid
Assay (GC)> 88.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only