Manganese powder, APS <10 micron, 99.6% (metals basis)
Manganese powder, APS <10 micron, 99.6% (metals basis)
Manganese powder, APS <10 micron, 99.6% (metals basis)
Thermo Scientific Chemicals

Manganese powder, APS <10 micron, 99.6% (metals basis)

CAS: 7439-96-5 | Mn | 54.94 g/mol
製品番号(カタログ番号) 045098.A1
または、製品番号045098-A1
価格(JPY)
-
数量:
1 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7439-96-5
IUPAC Namemanganese
Molecular FormulaMn
InChI KeyPWHULOQIROXLJO-UHFFFAOYSA-N
SMILES[Mn]
さらに表示
Total Metal Impurities0.4% max.
Appearance (Color)Gray
Particle SizeAPS < 10 microns
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only