Molybdenum gauze, 40 mesh woven from 0.13mm (0.005in) dia wire
Molybdenum gauze, 40 mesh woven from 0.13mm (0.005in) dia wire
Molybdenum gauze, 40 mesh woven from 0.13mm (0.005in) dia wire
Thermo Scientific Chemicals

Molybdenum gauze, 40 mesh woven from 0.13mm (0.005in) dia wire

CAS: 7439-98-7 | Mo | 95.95 g/mol
数量:
75 x 75 mm
150 x 150 mm
製品番号(カタログ番号) 046606.FL
または、製品番号046606-FL
価格(JPY)
-
数量:
75 x 75 mm
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
さらに表示
FormGauze, 40 mesh woven from 0.13 mm (0.005 in) dia. wire
CompositionMolybdenum: ≥99.9%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only