Tetraethyltin, 98%
Tetraethyltin, 98%
Tetraethyltin, 98%
Thermo Scientific Chemicals

Tetraethyltin, 98%

CAS: 597-64-8 | C8H20Sn | 234.96 g/mol
製品番号(カタログ番号) 071144.09
または、製品番号071144-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS597-64-8
IUPAC Nametetraethylstannane
Molecular FormulaC8H20Sn
InChI KeyRWWNQEOPUOCKGR-UHFFFAOYSA-N
SMILESCC[Sn](CC)(CC)CC
さらに表示
Assay (GC)> 97.0%
Tetraethyltin is used in the electronics industry. Used as catalyst for olefin polymerization and to produce other organotin compounds. Also used as a preservative for wood, textiles, paper, and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only