Titanium(IV) 2-ethylhexyloxide, 95%
Titanium(IV) 2-ethylhexyloxide, 95%
Titanium(IV) 2-ethylhexyloxide, 95%
Thermo Scientific Chemicals

Titanium(IV) 2-ethylhexyloxide, 95%

CAS: 1070-10-6 | C32H68O4Ti | 564.76 g/mol
数量:
100 g
500 g
2 kg
製品番号(カタログ番号) 077107.36
または、製品番号077107-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1070-10-6
IUPAC Nametitanium(4+) tetrakis(2-ethylhexan-1-olate)
Molecular FormulaC32H68O4Ti
InChI KeyKTXWGMUMDPYXNN-UHFFFAOYNA-N
SMILES[Ti+4].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-].CCCCC(CC)C[O-]
さらに表示
FormLiquid or viscous liquid
Color (APHA)≤100
Assay from Suppliers CofATitanium (Ti): 8.0-9.0%
Chloride content≤100ppm (0.10%)
Appearance (Color)Clear yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only