2-Biphenylcarboxylic acid, 98%
2-Biphenylcarboxylic acid, 98%
2-Biphenylcarboxylic acid, 98%
Thermo Scientific Chemicals

2-Biphenylcarboxylic acid, 98%

CAS: 947-84-2 | C13H10O2 | 198.22 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 106271000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS947-84-2
IUPAC Name[1,1'-biphenyl]-2-carboxylic acid
Molecular FormulaC13H10O2
InChI KeyILYSAKHOYBPSPC-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1C1=CC=CC=C1
さらに表示
Appearance (Color)White to beige
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Melting point110°C to 114°C
Appearance (Form)Crystalline powder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only