4-Bromochlorobenzene, 99%
4-Bromochlorobenzene, 99%
4-Bromochlorobenzene, 99%
Thermo Scientific Chemicals

4-Bromochlorobenzene, 99%

CAS: 106-39-8 | C6H4BrCl | 191.45 g/mol
数量:
50 g
500 g
1 kg
パッケージ:
Plastic bottle
製品番号(カタログ番号) 106830010
価格(JPY)
-
数量:
1 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS106-39-8
IUPAC Name1-bromo-4-chlorobenzene
Molecular FormulaC6H4BrCl
InChI KeyNHDODQWIKUYWMW-UHFFFAOYSA-N
SMILESClC1=CC=C(Br)C=C1
さらに表示
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point64°C to 67°C
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only