Cinnamonitrile, 97%, predominantly trans
Cinnamonitrile, 97%, predominantly trans
Cinnamonitrile, 97%, predominantly trans
Thermo Scientific Chemicals

Cinnamonitrile, 97%, predominantly trans

CAS: 1885-38-7 | C9H7N | 129.16 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 110380250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1885-38-7
IUPAC Name(2E)-3-phenylprop-2-enenitrile
Molecular FormulaC9H7N
InChI KeyZWKNLRXFUTWSOY-QPJJXVBHSA-N
SMILESN#C\C=C\C1=CC=CC=C1
さらに表示
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
Refractive index1.5990 to 1.6040 (20°C, 589 nm)
GC=<10 % (cis-Cinnamonitrile)
GC>=96.0 % (Sum of cis/trans)
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only