2,6-Dichloroaniline, 98%
2,6-Dichloroaniline, 98%
2,6-Dichloroaniline, 98%
Thermo Scientific Chemicals

2,6-Dichloroaniline, 98%

CAS: 608-31-1 | C6H5Cl2N | 162.01 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 113101000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS608-31-1
IUPAC Name2,6-dichloroaniline
Molecular FormulaC6H5Cl2N
InChI KeyJDMFXJULNGEPOI-UHFFFAOYSA-N
SMILESNC1=C(Cl)C=CC=C1Cl
さらに表示
Appearance (Color)White to brown
Melting point35°C to 39°C
GC>=97.5 %
Appearance (Form)Crystalline solid
AppearanceMay darken on storage
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only