Diethyl chlorophosphite, 90%, tech., Thermo Scientific Chemicals
Diethyl chlorophosphite, 90%, tech., Thermo Scientific Chemicals
Diethyl chlorophosphite, 90%, tech., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diethyl chlorophosphite, 90%, tech., Thermo Scientific Chemicals

数量:
25 mL
100 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 114250250
価格(JPY)
-
数量:
25 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS589-57-1
IUPAC Namediethyl chlorophosphonite
Molecular FormulaC4H10ClO2P
InChI KeyTXHWYSOQHNMOOU-UHFFFAOYSA-N
SMILESCCOP(Cl)OCC
さらに表示
Appearance (Form)Liquid
Titration Argentometric>=88 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4340 to 1.4410 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only