5,5-Dimethylhydantoin, 97%
5,5-Dimethylhydantoin, 97%
5,5-Dimethylhydantoin, 97%
Thermo Scientific Chemicals

5,5-Dimethylhydantoin, 97%

CAS: 77-71-4 | C5H8N2O2 | 128.13 g/mol
数量:
250 g
1 kg
5 kg
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 116310051
価格(JPY)
-
数量:
5 kg
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS77-71-4
Appearance (Color)White to almost white
Infrared spectrumConforms
Loss on drying=<0.5 % (60°C, 2 h, 1 mmHg)
Appearance (Form)Crystalline powder or crystals
Melting point175°C to 180°C
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only