Ethyl carbazate, 97%
Ethyl carbazate, 97%
Ethyl carbazate, 97%
Thermo Scientific Chemicals

Ethyl carbazate, 97%

CAS: 4114-31-2 | C3H8N2O2 | 104.11 g/mol
数量:
25 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 118200250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4114-31-2
IUPAC Nameethoxycarbohydrazide
Molecular FormulaC3H8N2O2
InChI KeyVYSYZMNJHYOXGN-UHFFFAOYSA-N
SMILESCCOC(=O)NN
さらに表示
Appearance (Color)May have a pink hue
Infrared spectrumConforms
Melting point42°C to 48°C
Appearance (Color)White to almost white
Appearance (Form)Crystals and/or chunks or fused solid
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only