Hexamethylenetetramine, 99%
Hexamethylenetetramine, 99%
Hexamethylenetetramine, 99%
Thermo Scientific Chemicals

Hexamethylenetetramine, 99%

CAS: 100-97-0 | C6H12N4 | 140.19 g/mol
数量:
1 kg
2.5 kg
10 kg
パッケージ:
すべて表示
Plastic Bottle
製品番号(カタログ番号) 120610010
価格(JPY)
-
数量:
1 kg
パッケージ:
Plastic Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS100-97-0
IUPAC Name1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
Molecular FormulaC6H12N4
InChI KeyVKYKSIONXSXAKP-UHFFFAOYSA-N
SMILESC1N2CN3CN1CN(C2)C3
さらに表示
Titration with HClO4>=98.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Ash=<0.1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only