4-Hydroxy-2-mercapto-6-methylpyrimidine, 98%, Thermo Scientific Chemicals
4-Hydroxy-2-mercapto-6-methylpyrimidine, 98%, Thermo Scientific Chemicals
4-Hydroxy-2-mercapto-6-methylpyrimidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxy-2-mercapto-6-methylpyrimidine, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量パッケージ
1213000252.5kgPlastic bottle
121305000500gPlastic bottle
製品番号(カタログ番号) 121300025
価格(JPY)
-
数量:
2.5kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS56-04-2
IUPAC Name6-methyl-2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
Molecular FormulaC5H6N2OS
InChI KeyHWGBHCRJGXAGEU-UHFFFAOYSA-N
SMILESCC1=CC(=O)NC(=S)N1
さらに表示
Appearance (Color)White to light yellow to beige
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only