Octadecylbenzene, 98%
Octadecylbenzene, 98%
Octadecylbenzene, 98%
Thermo Scientific Chemicals

Octadecylbenzene, 98%

CAS: 4445-07-2 | C24H42 | 330.59 g/mol
数量:
1 g
10 g
50 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 124700010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4445-07-2
IUPAC Nameoctadecylbenzene
Molecular FormulaC24H42
InChI KeyWSVDSBZMYJJMSB-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCC1=CC=CC=C1
さらに表示
Appearance (Form)Low melting solid
Melting point32°C to 36°C
Appearance (Color)White to light yellow
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only