4-Methylbenzophenone, 97%
4-Methylbenzophenone, 97%
4-Methylbenzophenone, 97%
Thermo Scientific Chemicals

4-Methylbenzophenone, 97%

CAS: 134-84-9 | C14H12O | 196.25 g/mol
数量:
50 g
250 g
パッケージ:
Plastic bottle
製品番号(カタログ番号) 126350500
価格(JPY)
-
数量:
50 g
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS134-84-9
IUPAC Name(4-methylphenyl)(phenyl)methanone
Molecular FormulaC14H12O
InChI KeyWXPWZZHELZEVPO-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
さらに表示
Appearance (Form)Powder or crystals or flakes or fused solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only