Methylene Blue trihydrate, 95%, pure
Methylene Blue trihydrate, 95%, pure
Methylene Blue trihydrate, 95%, pure
Thermo Scientific Chemicals

Methylene Blue trihydrate, 95%, pure

CAS: 7220-79-3 | C16H24ClN3O3S | 373.90 g/mol
数量:
100 g
250 g
1 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 126751000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7220-79-3
IUPAC Name3,7-bis(dimethylamino)-5λ⁴-phenothiazin-5-ylium trihydrate chloride
Molecular FormulaC16H24ClN3O3S
InChI KeyXQAXGZLFSSPBMK-UHFFFAOYSA-M
SMILESO.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
さらに表示
Appearance (Color)Dark green
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Loss on drying=<15 % (105°C)
Titration Argentometric>=94 % (on dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only