4-Methyl-2-nitrophenol, 99%
4-Methyl-2-nitrophenol, 99%
4-Methyl-2-nitrophenol, 99%
Thermo Scientific Chemicals

4-Methyl-2-nitrophenol, 99%

CAS: 119-33-5 | C7H7NO3 | 153.14 g/mol
数量:
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 127311000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS119-33-5
IUPAC Name4-methyl-2-nitrophenol
Molecular FormulaC7H7NO3
InChI KeySYDNSSSQVSOXTN-UHFFFAOYSA-N
SMILESCC1=CC=C(O)C(=C1)[N+]([O-])=O
さらに表示
Appearance (Form)Low melting crystalline mass
GC>=98.5 %
Melting point30°C to 34°C
Appearance (Color)Yellow to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only