3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%
3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%
3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%
Thermo Scientific Chemicals

3-Methyl-1-phenyl-2-pyrazolin-5-one, 99%

CAS: 89-25-8 | C10H10N2O | 174.2 g/mol
数量:
5 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 127461000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS89-25-8
IUPAC Name3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Molecular FormulaC10H10N2O
InChI KeyQELUYTUMUWHWMC-UHFFFAOYSA-N
SMILESCC1=NN(C(=O)C1)C1=CC=CC=C1
さらに表示
Infrared spectrumConforms
Melting point127°C to 131°C
Appearance (Form)Crystalline powder
HPLC>=98.5 %
Appearance (Color)Yellow to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only